Anti-PDGFRA Rabbit Polyclonal Antibody

Supplier: ProSci

70-646
10075-118EA 1042.19 CAD
10075-118
Anti-PDGFRA Rabbit Polyclonal Antibody
Antibodies
PDGFRA Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, Immunoprecipitation and Western Blotting starting at 1:250 - 1:1000.

0.2 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDGFRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Gibbon, Bovine, Chicken, Dog, Xenopus, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR