Anti-CLDN11 Rabbit Polyclonal Antibody

Supplier: ProSci
70-741
10075-146EA 1078.68 CAD
10075-146
Anti-CLDN11 Rabbit Polyclonal Antibody
Antibodies
CLDN11 / Claudin 11 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CLDN11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR