Anti-TNFRSF1B Rabbit Polyclonal Antibody

Supplier: ProSci

70-742
10075-148EA 1078.68 CAD
10075-148
Anti-TNFRSF1B Rabbit Polyclonal Antibody
Antibodies
TNFRSF1B / TNFR2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50, Immunoprecipitation and Western Blotting starting at 1 - 2?g/mL.

10 mM PBS, pH 7.4, BSA, with less than 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNFRSF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR