Anti-CD40 Rabbit Polyclonal Antibody

Supplier: ProSci
70-754
10075-172EA 983.75 CAD
10075-172
Anti-CD40 Rabbit Polyclonal Antibody
Antibodies
  • CD40 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, Immunoprecipitation and Western Blotting.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR