Anti-GJA1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-765
10074-050EA 758.96 CAD
10074-050
Anti-GJA1 Rabbit Polyclonal Antibody
GJA1 / Connexin 43 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:25, and Western Blotting starting at 2 - 4 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR