Anti-APP Rabbit Polyclonal Antibody

Supplier: ProSci
70-777
10080-214EA 758.96 CAD
10080-214
Anti-APP Rabbit Polyclonal Antibody
  • APP Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50, and Western Blotting starting at 1 μg/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR