Anti-INHA Rabbit Polyclonal Antibody

Supplier: ProSci
70-789
10080-238EA 1078.68 CAD
10080-238
Anti-INHA Rabbit Polyclonal Antibody
Antibodies
INHA / Inhibin Alpha Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: INHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR