Anti-CALB1 Rabbit Polyclonal Antibody

Supplier: ProSci

70-799
10080-258EA 1042.19 CAD
10080-258
Anti-CALB1 Rabbit Polyclonal Antibody
Antibodies
CALB1 / Calbindin Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1 - 2 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CALB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR