Anti-MKI67 Rabbit Polyclonal Antibody

Supplier: ProSci

70-805
10080-270EA 1042.19 CAD
10080-270
Anti-MKI67 Rabbit Polyclonal Antibody
Antibodies
MKI67 / Ki-67 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

10mM PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium Azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MKI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR