Anti-TGM2 Rabbit Polyclonal Antibody

Supplier: ProSci

70-808
10080-274EA 983.75 CAD
10080-274
Anti-TGM2 Rabbit Polyclonal Antibody
Antibodies
TGM2 / Transglutaminase 2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, Immunoprecipitation and Western Blotting starting at 5 - 10?g/mL.

10mM PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium Azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TGM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR