Anti-XIAP Rabbit Polyclonal Antibody

Supplier: ProSci

70-833
10075-188EA 1042.19 CAD
10075-188
Anti-XIAP Rabbit Polyclonal Antibody
Antibodies
XIAP Antibody can be used for Immunocytochemistry starting at 1:100 - 1:200, Immunofluorescence starting at 1:100 - 1:200, Immunohistochemistry in paraffin embeded tissues starting at 10?g/mL, and Western Blotting starting at 1:500 - 1:1000.

0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR