Anti-PYCARD Rabbit Polyclonal Antibody

Supplier: ProSci
70-867
10080-344EA 983.75 CAD
10080-344
Anti-PYCARD Rabbit Polyclonal Antibody
Antibodies
PYCARD Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1 - 2 ?g/mL.

0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PYCARD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR