Anti-NEIL3 Rabbit Polyclonal Antibody

Supplier: ProSci
70-871
10074-062EA 1042.19 CAD
10074-062
Anti-NEIL3 Rabbit Polyclonal Antibody
Antibodies
NEIL3 Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:500.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NEIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR