Anti-GP1BB Rabbit Polyclonal Antibody

Supplier: ProSci
70-881
10074-082EA 1042.19 CAD
10074-082
Anti-GP1BB Rabbit Polyclonal Antibody
Antibodies
GP1BB / CD42c Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:500 - 1:3000.

0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GP1BB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR