Anti-APEX1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-888
10074-096EA 983.75 CAD
10074-096
Anti-APEX1 Rabbit Polyclonal Antibody
Antibodies
  • APEX1 / APE1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting.

no preservative

Antibody should be stored at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: APEX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR