Anti-Red Blood Cells Rabbit Polyclonal Antibody

Supplier: ProSci

70-893
10080-354EA 1042.19 CAD
10080-354
Anti-Red Blood Cells Rabbit Polyclonal Antibody
Antibodies
Red Blood Cells Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100.

PBS, pH 7.2, 40% glycerol. No preservative added. No stabilizing proteins added.

Antibody should be stored at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: Red Blood Cells Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR