Anti-PARK7 Rabbit Polyclonal Antibody

Supplier: ProSci
70-894
10080-356EA 1221.42 CAD
10080-356
Anti-PARK7 Rabbit Polyclonal Antibody
Antibodies
PARK7 / DJ-1 Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:500 - 1:1000.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PARK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR