Anti-SUMO2 + SUMO3 Rabbit Polyclonal Antibody

Supplier: ProSci

70-898
10080-364EA 1042.19 CAD
10080-364
Anti-SUMO2 + SUMO3 Rabbit Polyclonal Antibody
Antibodies
SLC29A1 / ENT1 Antibody can be used for ELISA starting at 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL,

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SUMO2 / SUMO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR