Anti-NOS2 Rabbit Polyclonal Antibody

Supplier: ProSci

70-908
10080-384EA 1042.19 CAD
10080-384
Anti-NOS2 Rabbit Polyclonal Antibody
Antibodies
Nitric Oxide Synthase Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NOS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR