Anti-PRMT1 Rabbit Polyclonal Antibody

Supplier: ProSci

70-925
10080-418EA 1042.19 CAD
10080-418
Anti-PRMT1 Rabbit Polyclonal Antibody
Antibodies
KCNC2 / Kv3.2 Antibody can be used for ELISA starting at 1:10000, and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL,

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR