Anti-TNFRSF1A Rabbit Polyclonal Antibody

Supplier: ProSci
70-941
10080-450EA 758.96 CAD
10080-450
Anti-TNFRSF1A Rabbit Polyclonal Antibody
TNFRSF1A / TNFR1 Antibody can be used for Immunohistochemistry starting at 0.9 ?g/mL, Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting.

Borate buffer saline, pH 8.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Drosophila, Xenopus, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR