Anti-NANOG Rabbit Polyclonal Antibody

Supplier: ProSci
70-947
10074-110EA 835.8 CAD
10074-110
Anti-NANOG Rabbit Polyclonal Antibody
NANOG Antibody can be used for ELISA starting at 1:1000, Immunofluorescence starting at 1:10 - 1:50, Immunohistochemistry in paraffin embeded tissues starting at 10?g/mL, and Western Blotting starting at 1:50 - 1:100.

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR