Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: ProSci

70-950
10074-116EA 1078.68 CAD
10074-116
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
STAT1 Antibody can be used for ELISA starting at 1:10000, and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL,

PBS, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR