Anti-CASP3 Rabbit Polyclonal Antibody

Supplier: ProSci

70-955
10074-126EA 1042.19 CAD
10074-126
Anti-CASP3 Rabbit Polyclonal Antibody
Antibodies
CASP3 / Caspase 3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:1000.

PBS, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR