Anti-BRAP Rabbit Polyclonal Antibody

Supplier: ProSci

70-956
10074-128EA 1042.19 CAD
10074-128
Anti-BRAP Rabbit Polyclonal Antibody
Antibodies
BRAP Antibody can be used for ELISA starting at 1:000 - 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR