Anti-PTPRR Rabbit Polyclonal Antibody

70-958
10074-132EA 639.14 CAD
10074-132
Anti-PTPRR Rabbit Polyclonal Antibody
PTPRR Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTPRR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR