Anti-ADGRB3 Rabbit Polyclonal Antibody

Supplier: ProSci

70-961
10074-138EA 983.75 CAD
10074-138
Anti-ADGRB3 Rabbit Polyclonal Antibody
Antibodies
BAI3 Antibody can be used for ELISA starting at 1:000 - 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 15 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BAI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Chicken, Dog, Xenopus, Gorilla, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR