Anti-ADGRB2 Rabbit Polyclonal Antibody

Supplier: ProSci
70-962
10074-140EA 758.96 CAD
10074-140
Anti-ADGRB2 Rabbit Polyclonal Antibody
BAI2 Antibody can be used for ELISA starting at 1:000 - 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADGRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR