Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: ProSci

70-973
10074-160EA 1042.19 CAD
10074-160
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
MMP9 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 2 - 4 ?g/mL.

10 mM PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea pig, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR