Anti-TRAF6 Rabbit Polyclonal Antibody

70-974
10074-162EA 758.96 CAD
10074-162
Anti-TRAF6 Rabbit Polyclonal Antibody
TRAF6 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50, Immunoprecipitation and Western Blotting starting at 1 - 3?g/mL.

PBS, pH 7.4, 0.1% sodium azide, before the addition of glycerol to 40%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRAF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR