Anti-ID3 Rabbit Polyclonal Antibody

Supplier: ProSci

70-985
10075-240EA 1078.68 CAD
10075-240
Anti-ID3 Rabbit Polyclonal Antibody
Antibodies
ID3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ID3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR