Anti-SOX9 Rabbit Polyclonal Antibody

Supplier: Prosci

70-992
10080-452EA 983.75 CAD
10080-452
Anti-SOX9 Rabbit Polyclonal Antibody
Antibodies
SOX9 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

10mM PBS/1% BSA buffer pH 7.4 with 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SOX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR