Anti-FCER2 Rabbit Polyclonal Antibody

71-013
10080-490EA 758.96 CAD
10080-490
Anti-FCER2 Rabbit Polyclonal Antibody
FCER2 / CD23 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 15 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FCER2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR