Anti-CTSD Rabbit Polyclonal Antibody

Supplier: Prosci

71-016
10080-496EA 1042.19 CAD
10080-496
Anti-CTSD Rabbit Polyclonal Antibody
Antibodies
CTSD / Cathepsin D Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR