Anti-CD44 Rabbit Polyclonal Antibody

71-027
10080-518EA 758.96 CAD
10080-518
Anti-CD44 Rabbit Polyclonal Antibody
CD44 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD44
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR