Anti-SFN Rabbit Polyclonal Antibody

Supplier: Prosci

71-028
10080-520EA 1157.94 CAD
10080-520
Anti-SFN Rabbit Polyclonal Antibody
Antibodies
SFN / 14-3-3 Sigma Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:250, and Western Blotting.

0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SFN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR