Anti-MAP3K1 Rabbit Polyclonal Antibody

71-031
10080-526EA 639.14 CAD
10080-526
Anti-MAP3K1 Rabbit Polyclonal Antibody
MAP3K1 / MEKK1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:1000.

25mM sodium borate, 100mM boric acid, pH 8.4, 75mM NaCl, 5mM EDTA

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAP3K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR