Anti-KLK3 Rabbit Polyclonal Antibody

71-037
10080-538EA 835.8 CAD
10080-538
Anti-KLK3 Rabbit Polyclonal Antibody
KLK3 / PSA Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR