Anti-A2M Rabbit Polyclonal Antibody

71-038
10080-540EA 1042.19 CAD
10080-540
Anti-A2M Rabbit Polyclonal Antibody
Antibodies
A2M / Alpha-2-Macroglobulin Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR