Anti-DDX3X Rabbit Polyclonal Antibody

Supplier: Prosci

71-039
10074-166EA 1042.19 CAD
10074-166
Anti-DDX3X Rabbit Polyclonal Antibody
Antibodies
DDX3X Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 20 ?g/mL, and Western Blotting starting at 1?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DDX3X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR