Anti-FOXL1 Rabbit Polyclonal Antibody

71-043
10074-172EA 1042.19 CAD
10074-172
Anti-FOXL1 Rabbit Polyclonal Antibody
Antibodies
FOXL1 Antibody can be used for ELISA starting at 1:200000, and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, sodium azide, before the addition of glycerol to 40%.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR