Anti-MKI67 Rabbit Polyclonal Antibody

71-052
10077-566EA 1042.19 CAD
10077-566
Anti-MKI67 Rabbit Polyclonal Antibody
Antibodies
MKI67 / Ki-67 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 40 ?g/mL.

20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MKI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR