Anti-LMNA Rabbit Polyclonal Antibody

Supplier: Prosci

71-055
10077-572EA 1078.68 CAD
10077-572
Anti-LMNA Rabbit Polyclonal Antibody
Antibodies
Lamin C Antibody can be used for Immunofluorescence, Immunohistochemistry in paraffin embeded tissues starting at 1:50, and Western Blotting.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LMNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR