Anti-CTNND1 Rabbit Polyclonal Antibody

Supplier: Prosci

71-060
10080-550EA 1042.19 CAD
10080-550
Anti-CTNND1 Rabbit Polyclonal Antibody
Antibodies
CTNND1 / p120CTN Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:15.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CTNND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR