Anti-RCC1 Rabbit Polyclonal Antibody

71-065
10080-558EA 1042.19 CAD
10080-558
Anti-RCC1 Rabbit Polyclonal Antibody
Antibodies
RCC1 Antibody can be used for Immunofluorescence, Immunohistochemistry in paraffin embeded tissues 1:100, Immunoprecipitation and Western Blotting starting at 1:500.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR