Anti-ALPP Rabbit Polyclonal Antibody

71-067
10080-562EA 758.96 CAD
10080-562
Anti-ALPP Rabbit Polyclonal Antibody
ALPP Antibody can be used for ELISA , Immunofluorescence starting at 1:100, Immunohistochemistry in paraffin embeded tissues starting at 5?g/mL, Immunoprecipitation starting at 1:100, and Western Blotting starting at 1:100.

10 mM KPO4, pH 6.4. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ALPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR