Anti-TH Rabbit Polyclonal Antibody

Supplier: Prosci

71-068
10080-564EA 1042.19 CAD
10080-564
Anti-TH Rabbit Polyclonal Antibody
Antibodies
TH / Tyrosine Hydroxylase Antibody can be used for Immunofluorescence starting at 1:1000, Immunohistochemistry starting at 1:1000, Immunohistochemistry in frozen tissues Immunohistochemistry in paraffin embeded tissues 1:100, and Western Blotting starting at1:1000.

50 uL of 10 mM HEPES, pH 7.5, 150 mM NaCl, 0.1 mg/mL BSA, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR