Anti-CDHR5 Rabbit Polyclonal Antibody

Supplier: Prosci

71-078
10075-256EA 1042.19 CAD
10075-256
Anti-CDHR5 Rabbit Polyclonal Antibody
Antibodies
CDHR5 / MUPCDH Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDHR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR