Anti-SRPX2 Rabbit Polyclonal Antibody

Supplier: Prosci

71-080
10080-580EA 1042.19 CAD
10080-580
Anti-SRPX2 Rabbit Polyclonal Antibody
Antibodies
SRPX2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 15 - 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SRPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Chimpanzee, Gorilla, Human, Orangutan, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR