To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:sushi-repeat containing protein, X-linked 2
- Clonality:Polyclonal
- Gene ID:O60687
- Host:Rabbit
- Reactivity:Guinea pig,Dog,Mouse,Bat,Pig,Rabbit,Horse,Monkey,Gibbon,Gorilla,Orangutan,Chimpanzee,Human,Bovine
- Antigen symbol:SRPX2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Antigen synonyms:SRPX2
- Storage buffer:PBS, 0.1% sodium azide.
- Storage temperature:Antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
- Concentration:0.5 mg/mL
- Shipping temperature:4 °C
- Immunogen:Internal domain of human
- Tested applications:Immunoaffinity Chromatography
- Purification:Immunoaffinity Chromatography
- Cat. no.:10080-582
Specifications
About this item
SRPX2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 15 ?g/mL.
PBS, 0.1% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SRPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chimpanzee, Dog, Guinea pig, Gorilla, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rabbit